Skip to Content
Merck
All Photos(1)

Key Documents

665169

Sigma-Aldrich

trans-1-Penten-1-ylboronic acid pinacol ester

97%

Synonym(s):

E-2-(1-Pentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-1-Pentenylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21BO2
CAS Number:
Molecular Weight:
196.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.441

bp

110-111 °C/28 mmHg

density

0.884 g/mL at 25 °C

SMILES string

CCC\C=C\B1OC(C)(C)C(C)(C)O1

InChI

1S/C11H21BO2/c1-6-7-8-9-12-13-10(2,3)11(4,5)14-12/h8-9H,6-7H2,1-5H3/b9-8+

InChI key

IMIHEZMTQOARIK-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service