Skip to Content
Merck
All Photos(1)

Documents

663506

Sigma-Aldrich

Erythromycin-(N-methyl-13C,d3)

99 atom % 13C, 98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC36D3H64NO13
Molecular Weight:
737.94
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

98% (CP)

form

solid

mol wt

737.87 by atom % calculation

technique(s)

mass spectrometry (MS): suitable

mp

133-135 °C

mass shift

M+4

SMILES string

[2H][13C]([2H])([2H])N(C)[C@H]1C[C@@H](C)O[C@@H](O[C@@H]2[C@@H](C)[C@H](O[C@H]3C[C@@](C)(OC)[C@@H](O)[C@H](C)O3)[C@@H](C)C(=O)O[C@H](CC)[C@@](C)(O)[C@H](O)[C@@H](C)C(=O)[C@H](C)C[C@@]2(C)O)[C@@H]1O

InChI

1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1/i11+1D3

InChI key

ULGZDMOVFRHVEP-ZZKXWFBHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kaleb J Duelge et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1053, 81-86 (2017-04-18)
Antibiotics are used in ethanol production to discourage the growth of bacteria that would result in lower ethanol content and a lower quality product. A survey conducted by the FDA (FY 2010 Nationwide Survey of Distillers Grains for Antibiotic Residues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service