Skip to Content
Merck
All Photos(2)

Key Documents

662909

Sigma-Aldrich

2-Pentynoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€330.00

€330.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€330.00

About This Item

Linear Formula:
CH3CH2C≡CCO2H
CAS Number:
Molecular Weight:
98.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€330.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

47-53 °C

functional group

carboxylic acid

SMILES string

CCC#CC(O)=O

InChI

1S/C5H6O2/c1-2-3-4-5(6)7/h2H2,1H3,(H,6,7)

InChI key

MINRDQDGBLQBGD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

205.0 °F

Flash Point(C)

96.11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Tiwari et al.
International journal of pharmaceutics, 531(1), 281-291 (2017-08-29)
Nanoparticles of cationized polyacrylamide (CPAA NP), decorated with the recognition peptide VRPMPLQ (to produce CPAA-Pep NP), were prepared, characterized and tested biologically. They were designed to target dysplasia regions in the colon, characterized by overexpressed sialic acid. This targetability was
S Shi et al.
Biosensors & bioelectronics, 72, 205-210 (2015-05-20)
We propose in this work a general and versatile methodology for electrochemical monitoring of persistent pharmaceutical micropollutants. The system presented is based on an electroactive and electropolymerized hapten (mimetic molecule of the pollutant to be detected) and a specific antibody

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service