Skip to Content
Merck
All Photos(1)

Documents

662135

Sigma-Aldrich

Nτ-Methyl-d3-L-histidine

98 atom % D, 99% (CP)

Synonym(s):

1-Methyl-d3-L-histidine, 3-Methyl-d3-L-histidine (archaic), tau-Methyl-d3-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D3H9N3O2
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]20/D -24°, c = 2 in H2O

mp

~240 °C (dec.)

mass shift

M+3

SMILES string

[H][C@](N)(Cc1cn(cn1)C([2H])([2H])[2H])C(O)=O

InChI

1S/C7H11N3O2/c1-10-3-5(9-4-10)2-6(8)7(11)12/h3-4,6H,2,8H2,1H3,(H,11,12)/t6-/m0/s1/i1D3

InChI key

BRMWTNUJHUMWMS-FYFSCIFKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service