Skip to Content
Merck
All Photos(2)

Key Documents

661023

Sigma-Aldrich

Hydroxy(cyclooctadiene)rhodium(I) dimer

greener alternative

95%

Synonym(s):

(Cyclooctadiene)rhodium hydroxide dimer, 1,5-Cyclooctadiene)hydroxyrhodium dimer, Bis(cyclooctadiene)dihydroxodirhodium, Bis[(1,2,5,6-η)-1,5-cyclooctadiene]di--μ-hydroxydirhodium, Bis[(1,5-cyclooctadiene)(hydroxy)rhodium], [Rh(OH)(1,5-cod)]2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€300.00
1 G
€333.00

€300.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€300.00
1 G
€333.00

About This Item

Empirical Formula (Hill Notation):
C16H26O2Rh2
CAS Number:
Molecular Weight:
456.19
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€300.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

199 °C (dec.)

greener alternative category

SMILES string

O[Rh].O[Rh].C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.2H2O.2Rh/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;2*1H2;;/q;;;;2*+1/p-2/b2*2-1-,8-7-;;;;

InChI key

IUAQXTGDGDIMOD-MIXQCLKLSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for oxygenative addition to terminal alkynes for a greener synthesis esters, amides, and carboxylic acids.

Rhodium-Catalyzed Oxygenative Addition to Terminal Alkynes for the Synthesis of Esters, Amides, and Carboxylic Acids
Rhodium catalyst employed with R-BINAP or R-SEGPHOS® ligands in a synthesis of chiral 3,3-disubstituted-1-indanones.[1]

Legal Information

SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-catalyzed asymmetric synthesis of 3,3-disubstituted 1-indanones.
Ryo Shintani et al.
Angewandte Chemie (International ed. in English), 46(20), 3735-3737 (2007-04-05)

Articles

Discover the role of boronic acids in enantioenriched C(sp2)–C(sp2) product synthesis, specifically the rhodium-catalyzed Suzuki–Miyaura-type arylations of allylic halides and cyclobutenes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service