Skip to Content
Merck
All Photos(1)

Key Documents

659223

Sigma-Aldrich

Poly(2,5-bis(3-sulfonatopropoxy)-1,4-phenylene, disodium salt-alt-1,4-phenylene)

light emitting, fluorescence λem 424 nm in H2O

Synonym(s):

(−)PPP

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€428.00

€428.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€428.00

About This Item

Linear Formula:
(C18H18Na2O8S2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€428.00


Check Cart for Availability

Request a Bulk Order

form

solid

solubility

H2O: soluble

λmax

208 nm (in deoxygenated water)(lit.)

fluorescence

λex 322 nm; λem 424 nm in H2O
λex 363 nm; λem 410 nm in H2O(lit.)
λem 424 nm in H2O

General description

Poly(2,5-bis(3-sulfonatopropoxy)-1,4-phenylene, disodium salt-alt-1,4-phenylene) (PPP-OPSO3) is a water soluble conjugating polymer that can be used as a donor molecule. It has a poly(p-phenylene) skeleton with each repeating unit having two phenylene groups. It can be used for a variety of optoelectronic applications.[1][2]

Application

A water-soluble polyanionic blue-light emitting polymer.
PPP-OPSO3 is a π-conjugating polymer that can be used in organic light emitting diodes (OLEDs), thin film transistors (TFTs), and flexible organic solar cells.[1][2][3]

Features and Benefits

Ready to use, water-soluble light, emitting material, more soluble than MPS-PPV.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled energy transfer between isolated donor-acceptor molecules intercalated in thermally self-ensemble two-dimensional hydrogen bonding cages
Al Attar HA and Monkman AP
Physical Review. B, Condensed Matter and Materials Physics, 86(23), 235420-235420 (2012)
Conjugated Polymer Nanoparticles Having Modified Band Gaps Assembled into Nano-and Micropatterned Organic Light-Emitting Diodes
Herrera M, et al.
ACS Applied Nano Materials, 2(1), 577-585 (2018)
Room-Temperature Phosphorescence From Films of Isolated Water-Soluble Conjugated Polymers in Hydrogen-Bonded Matrices
Al-Attar HA and Monkman AP
Advances in Functional Materials, 22(18), 3824-3832 (2012)
Reynolds, J.R., et al.
Macromolecules, 31, 964-964 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service