Skip to Content
Merck
All Photos(1)

Key Documents

656712

Sigma-Aldrich

(R)-(+)-3-(Dimethylamino)pyrrolidine

95%

Synonym(s):

(3R)-N,N-Dimethyl-3-pyrrolidinamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€101.00

€101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€101.00

About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€101.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

optical activity

[α]20/D +14°, c = 1 in ethanol

refractive index

n20/D 1.4650 (lit.)

bp

166 °C/760 mmHg (lit.)

density

0.899 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)[C@@H]1CCNC1

InChI

1S/C6H14N2/c1-8(2)6-3-4-7-5-6/h6-7H,3-5H2,1-2H3/t6-/m1/s1

InChI key

AVAWMINJNRAQFS-ZCFIWIBFSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.1 °F - (External MSDS)

Flash Point(C)

51.7 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service