Skip to Content
Merck

Skip To

656151

3-(4-Chlorophenyl)propionic acid

97%

Synonym(s):

p-Chlorohydrocinnamic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
€ 146,00

About This Item

Linear Formula:
ClC6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
184.62
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
97%
Form:
solid

€ 146,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

solid

mp

127-131 °C (lit.)

functional group

carboxylic acid, chloro

SMILES string

OC(=O)CCc1ccc(Cl)cc1

InChI

1S/C9H9ClO2/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-2,4-5H,3,6H2,(H,11,12)

InChI key

BBSLOKZINKEUCR-UHFFFAOYSA-N



Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters