Skip to Content
Merck
All Photos(1)

Key Documents

656011

Sigma-Aldrich

2,4-Dichloro-6-(trifluoromethyl)aniline

97%

Synonym(s):

2-Amino-3,5-dichlorobenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CL2C6H2(CF3)NH2
CAS Number:
Molecular Weight:
230.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.522

bp

98-100 °C/0.6 mmHg

density

1.532 g/mL at 25 °C

functional group

chloro
fluoro

SMILES string

Nc1c(Cl)cc(Cl)cc1C(F)(F)F

InChI

1S/C7H4Cl2F3N/c8-3-1-4(7(10,11)12)6(13)5(9)2-3/h1-2H,13H2

InChI key

OCJPQZFOBGQYEA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service