Skip to Content
Merck
All Photos(1)

Key Documents

655147

Sigma-Aldrich

2,4,6-Trichloroanisole-d5

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€446.00
500 MG
€3,680.00

€446.00


Check Cart for Availability

Request more information

Select a Size

Change View
50 MG
€446.00
500 MG
€3,680.00

About This Item

Linear Formula:
Cl3C6D2OCD3
CAS Number:
Molecular Weight:
216.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

€446.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

bp

132 °C/28 mmHg (lit.)

mp

60-62 °C (lit.)

format

neat

mass shift

M+5

SMILES string

[2H]c1c(Cl)c([2H])c(Cl)c(OC([2H])([2H])[2H])c1Cl

InChI

1S/C7H5Cl3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3/i1D3,2D,3D

InChI key

WCVOGSZTONGSQY-RHIBPKLGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

This product may be used as an analytical standard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Navideh Sadoughi et al.
Journal of agricultural and food chemistry, 63(11), 2877-2885 (2015-02-24)
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service