Skip to Content
Merck
All Photos(1)

Key Documents

653667

Sigma-Aldrich

Pd(0) EnCat® 30NP

wet, nanoparticles, extent of labeling: 0.4 mmol/g Pd(0) loading (dry basis)

Synonym(s):

Palladium(0), microencapsulated in polyurea matrix

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€289.00

€289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€289.00

About This Item

Empirical Formula (Hill Notation):
Pd
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€289.00


Check Cart for Availability

Request a Bulk Order

form

nanoparticles
wet

Quality Level

contains

0.35-0.43 mmol/g pallladium(0) (dry basis)
45% water

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

0.4 mmol/g Pd(0) loading (dry basis)

weight

Unit weight includes weight of water.

General description

Pd(0) EnCat® 30NP may be used as a catalyst for the selective reduction of aromatic aldehydes, ketones and epoxides to form the corresponding alcohols via hydrogenation.

Other Notes

Encapsulated Pd catalyst reduction of aryl ketones and benzyl ethers.[1]
Matrix content 30%

Legal Information

Pd EnCat® technology.
EnCat is a registered trademark of Reaxa Ltd.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-heat. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Steven V Ley et al.
Organic letters, 5(24), 4665-4668 (2003-11-25)
[reaction: see text] Pd nanoparticles (approximately 2 nm in size) microencapsulated in polyurea is an efficient and recyclable catalyst for reductive ring-opening hydrogenolysis of epoxides, using either HCOOH/Et(3)N or H(2) as a hydrogen donor.

Related Content

Homogeneous palladium catalysts are widely utilized due to their versatility, reactivity and functional group tolerance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service