Skip to Content
Merck
All Photos(2)

Key Documents

652008

Sigma-Aldrich

4-Fluorophenethyl bromide

97%

Synonym(s):

1-Fluoro-4-(2-bromoethyl)benzene, 4-Fluoro-1-(2-bromoethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH2Br
CAS Number:
Molecular Weight:
203.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.534 (lit.)

bp

100-104 °C/15 mmHg (lit.)

density

1.4498 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(CCBr)cc1

InChI

1S/C8H8BrF/c9-6-5-7-1-3-8(10)4-2-7/h1-4H,5-6H2

InChI key

FLRUNCJXOVYWDH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Son T Nguyen et al.
Bioorganic & medicinal chemistry, 23(9), 2024-2034 (2015-03-31)
Recently we described a novel pyranopyridine inhibitor (MBX2319) of RND-type efflux pumps of the Enterobacteriaceae. MBX2319 (3,3-dimethyl-5-cyano-8-morpholino-6-(phenethylthio)-3,4-dihydro-1H-pyrano[3,4-c]pyridine) is structurally distinct from other known Gram-negative efflux pump inhibitors (EPIs), such as 1-(1-naphthylmethyl)-piperazine (NMP), phenylalanylarginine-β-naphthylamide (PAβN), D13-9001, and the pyridopyrimidine derivatives. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service