Skip to Content
Merck
All Photos(1)

Documents

651753

Sigma-Aldrich

2-Chloro-5-fluoropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2ClFN2
CAS Number:
Molecular Weight:
132.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.503 (lit.)

density

1.439 g/mL at 20 °C (lit.)

SMILES string

Fc1cnc(Cl)nc1

InChI

1S/C4H2ClFN2/c5-4-7-1-3(6)2-8-4/h1-2H

InChI key

AGYUQBNABXVWMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-5-fluoropyrimidine can be used as a starting material for the preparation of:
  • 5-fluoro-2-amino pyrimidines by reacting with various amines in the presence of K2CO3, via C-N bond forming reaction.
  • 2-chloro-5-(5-fluoropyrimidin-2-yl)benzoic acid, an intermediate, which is used in the synthesis of benzamide scaffolds as potent antagonists against P2X7 receptors.
  • 5-fluoro-2-cyano pyrimidine, a key intermediate used for the synthesis of 5-chloro-N-[(1S)-1-(5-fluoropyrimidin-2-yl)ethyl]-N-(5-methyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine, as a potent inhibitor of the JAK2 kinase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-free site-selective C-N bond-forming reaction of polyhalogenated pyridines and pyrimidines
Wang L, et al.
Royal Society of Chemistry Advances, 5(100), 82097-82111 (2015)
Discovery of 2-chloro-N-((4, 4-difluoro-1-hydroxycyclohexyl) methyl)-5-(5-fluoropyrimidin-2-yl) benzamide as a potent and CNS penetrable P2X7 receptor antagonist
Chen X, et al.
Bioorganic & Medicinal Chemistry Letters, 20(10), 3107-3111 (2010)
Use of ?-transaminase enzyme chemistry in the synthesis of a JAK2 kinase inhibitor
Frodsham L, et al.
Organic Process Research & Development, 17(9), 1123-1130 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service