Skip to Content
Merck
All Photos(1)

Key Documents

649821

Sigma-Aldrich

PEDOT

tetramethacrylate end-capped solution, 0.5 wt. % (dispersion in nitromethane), contains p-toluenesulfonate as dopant

Synonym(s):

Oligotron NM, PEDOT, PEDOT, tetramethacrylate end-capped

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€136.00

€136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€136.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€136.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(3,4-ethylenedioxythiophene), tetramethacrylate end-capped solution, 0.5 wt. % (dispersion in nitromethane), contains p-toluenesulfonate as dopant

mol wt

~1,360-1,600 g/mol (methacrylate equivalent weight)
average Mn ~6,000

Quality Level

contains

p-toluenesulfonate as dopant

composition

ethanol, 5.8 wt. %
isopropanol, 0.3 wt. %
nitromethane, 93.4 wt. %
Oligotron tetramethacrylate, 0.5 wt. %

concentration

0.5 wt. % (dispersion in nitromethane)

resistance

1-10 M Ω/sq (surface resistance of film)

conductivity

0.1-0.5 S/cm (bulk)

density

1.127 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Please follow this link for Aedotron/Oligotron selection guide table.

Application

For best results, agitate for 20 minutes prior to use. Spin coat as-is at 500 RPM for 9 seconds followed by 1500 RPM for 15 seconds. Photocurable inks can be formulated by adding acrylate monomers and photoinitiators (i.e.DMPA) to the nitromethane dispersion. The solvent must be evaporated prior to crosslinking. Functional compounds can be coupled to the oligomer via reactions at the methacrylate group.
Poly(3,4-ethylenedioxythiophene), tetramethacrylate end-capped solution (PEDOT-TMA) can be used as a hole injection layer (HIL) which can be used in optoelectronic applications such as light emitting diodes (LEDs), biosensors, and energy storage devices.[1][2]
Suitable for spin coating applications.
Used for depositing thin films of conducting EDOT oligomers for sensors, antistatic layers, organic solar cells and flexible electronics.

Features and Benefits

End-capped structure renders the conducting oligomer PEDOT dispersible. The methacrylate groups offer the potential for crosslinking, or additional chemical functionalization. Nitromethane has a suitable volatility for spin coating applications.

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Legal Information

Oligotron is a trademark of TDA Research, Inc.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly aligned carbon nanotube/polymer composites with much improved electrical conductivities
Peng H and Sun X
Chemical Physics Letters, 471(1-3), 103-105 (2009)
Cristina Ocaña et al.
Talanta, 186, 279-285 (2018-05-23)
We report here the fabrication of solid-contact calcium-selective electrodes (Ca2+-SCISEs) made of a polyurethane acrylate ion-selective membrane (ISM) that was covalently attached to the underlying ion-to-electron transducer (solid-contact). Methacrylate-functionalized poly(3,4-ethylenedioxythiophene) (Meth-PEDOT) and Meth-PEDOT films containing either multiwalled carbon nanotubes (MWCNT)

Articles

Conjugated polymers offer charge transport between inorganic, electrically conducting metals and organic, proton-conducting biological systems.

Progress in Organic Thermoelectric Materials & Devices including high ZT values of >0.2 at room temperature by p-type (PEDOT:PSS) & n-type (Poly[Kx(Ni-ett)]) materials are discussed.

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service