Skip to Content
Merck
All Photos(1)

Key Documents

649511

Sigma-Aldrich

9-(2-Ethylhexyl)carbazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€84.70

€84.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€84.70

About This Item

Empirical Formula (Hill Notation):
C20H25N
CAS Number:
Molecular Weight:
279.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€84.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5949 (lit.)

bp

40 °C/3 mmHg (lit.)

density

1.004 g/mL at 25 °C (lit.)

OLED Device Performance

ITO/PEDOT:PSS/EHCz:Rubrene (1 wt %)/Cs2CO3:ITO

  • Color: red
  • Max. EQE: 0.03 %

SMILES string

CCCCC(CC)Cn1c2ccccc2c3ccccc13

InChI

1S/C20H25N/c1-3-5-10-16(4-2)15-21-19-13-8-6-11-17(19)18-12-7-9-14-20(18)21/h6-9,11-14,16H,3-5,10,15H2,1-2H3

InChI key

SAXNQWFLHXTRDI-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic light-emitting diode with liquid emitting layer
Xu, D.; et al.
Applied Physics Letters, 95, 053304-053304 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service