Skip to Content
Merck
All Photos(1)

Key Documents

648469

Sigma-Aldrich

Methyl 5-(4-aminophenyl)furan-2-carboxylate

97%

Synonym(s):

5-(4-Aminophenyl)furan-2-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

122-126 °C (lit.)

SMILES string

COC(=O)c1ccc(o1)-c2ccc(N)cc2

InChI

1S/C12H11NO3/c1-15-12(14)11-7-6-10(16-11)8-2-4-9(13)5-3-8/h2-7H,13H2,1H3

InChI key

JOIQXOMSFVFDQY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María C M Aboy-Pardal et al.
Computational and structural biotechnology journal, 21, 224-237 (2022-12-23)
Caveolae are nanoscopic and mechanosensitive invaginations of the plasma membrane, essential for adipocyte biology. Transmission electron microscopy (TEM) offers the highest resolution for caveolae visualization, but provides complicated images that are difficult to classify or segment using traditional automated algorithms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service