Skip to Content
Merck
All Photos(1)

Key Documents

646229

Sigma-Aldrich

2′-Deoxycytidine-13C9,15N3 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)

Synonym(s):

13C and 15N Labeled 2′deoxycytidine 5′-triphosphate disodium salt solution, dCTP-13C9,15N3 disodium salt solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€121.00
10 MG
€515.00
25 MG
€1,120.00

€121.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
€121.00
10 MG
€515.00
25 MG
€1,120.00

About This Item

Empirical Formula (Hill Notation):
13C9H1415N3Na2O13P3
Molecular Weight:
523.03
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.12

€121.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom %

Assay

≥95% (CP)

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+12

shipped in

dry ice

storage temp.

−20°C

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service