Skip to Content
Merck
All Photos(1)

Key Documents

644110

Sigma-Aldrich

N-(2,3-Epoxypropyl)phthalimide

≥95.0%

Synonym(s):

N-Glycidylphthalimide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€169.00

€169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€169.00

About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
Beilstein:
171277
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€169.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0%

form

solid

mp

92-98 °C (lit.)

SMILES string

O=C1N(CC2CO2)C(=O)c3ccccc13

InChI

1S/C11H9NO3/c13-10-8-3-1-2-4-9(8)11(14)12(10)5-7-6-15-7/h1-4,7H,5-6H2

InChI key

DUILGEYLVHGSEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service