Skip to Content
Merck
All Photos(1)

Key Documents

643793

Sigma-Aldrich

2-Ethyl-4-methyl-1H-imidazole-1-propanenitrile

85% (sum of isomers)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€33.10

€33.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€33.10

About This Item

Empirical Formula (Hill Notation):
C9H13N3
CAS Number:
Molecular Weight:
163.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€33.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

85% (sum of isomers)

refractive index

n20/D 1.5070 (lit.)

bp

245-246 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

CCc1nc(C)cn1CCC#N

InChI

1S/C9H13N3/c1-3-9-11-8(2)7-12(9)6-4-5-10/h7H,3-4,6H2,1-2H3

InChI key

UIDDPPKZYZTEGS-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service