Skip to Content
Merck
All Photos(1)

Documents

643033

Sigma-Aldrich

4-Chloro-3-(trifluoromethyl)benzylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3CC6H3(Cl)CH2NH2
CAS Number:
Molecular Weight:
209.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.495

density

1.379 g/mL at 25 °C

SMILES string

NCc1ccc(Cl)c(c1)C(F)(F)F

InChI

1S/C8H7ClF3N/c9-7-2-1-5(4-13)3-6(7)8(10,11)12/h1-3H,4,13H2

InChI key

SRHQOQMNBVOXCF-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon H Rüdisser et al.
Journal of biomolecular NMR, 74(10-11), 579-594 (2020-06-20)
Fluorine NMR has recently gained high popularity in drug discovery as it allows efficient and sensitive screening of large numbers of ligands. However, the positive hits found in screening must subsequently be ranked according to their affinity in order to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service