Skip to Content
Merck
All Photos(3)

Key Documents

641359

Sigma-Aldrich

5-Bromo-2-pyridinecarbonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€37.90

€37.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€37.90

About This Item

Empirical Formula (Hill Notation):
C6H3BrN2
CAS Number:
Molecular Weight:
183.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€37.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

100-110 °C/3 mmHg (lit.)

mp

128-132 °C (lit.)

functional group

bromo
nitrile

SMILES string

Brc1ccc(nc1)C#N

InChI

1S/C6H3BrN2/c7-5-1-2-6(3-8)9-4-5/h1-2,4H

InChI key

DMSHUVBQFSNBBL-UHFFFAOYSA-N

Application

5-Bromo-2-pyridinecarbonitrile can be used in the synthesis of aza-terphenyl diamidine analogs, which exhibits potent antiprotozoal activity.[1] It can also be used in the synthesis of pyridine-diketopyrrolopyrrole(PyDPP), a building block for preparing low band-gap copolymers for use as electron donor in polymer solar cells.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis and activity of azaterphenyl diamidines against Trypanosoma brucei rhodesiense and Plasmodium falciparum"
Hu L, et al.
Bioorganic & Medicinal Chemistry, 17(18), 6651-6658 (2009)
"Synthesis of pyridine-capped diketopyrrolopyrrole and its use as a building block of low band-gap polymers for efficient polymer solar cells"
Jung WJ, et al.
Chemical Communications (Cambridge, England), 49(76), 8495-8497 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service