Skip to Content
Merck
All Photos(1)

Key Documents

640360

Sigma-Aldrich

Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I)tetrafluoroborate

Synonym(s):

[Rh(cod)(MeCN)2]BF4

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€478.00
5 G
€2,290.00

€478.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€478.00
5 G
€2,290.00

About This Item

Empirical Formula (Hill Notation):
C12H18BF4N2Rh
CAS Number:
Molecular Weight:
379.99
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€478.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

SMILES string

[Rh+].CC#N.CC#N.F[B-](F)(F)F.C1CC=CCCC=C1

InChI

1S/C8H12.2C2H3N.BF4.Rh/c1-2-4-6-8-7-5-3-1;2*1-2-3;2-1(3,4)5;/h1-2,7-8H,3-6H2;2*1H3;;/q;;;-1;+1/b2-1-,8-7-;;;;

InChI key

KSMAFOANQHYEJG-SUESZSCISA-N

Application

Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I)tetrafluoroborate [Rh(cod)(MeCN)2]BF4 may be used as a catalyst for the rhodium(I)-catalyzed silylation of aryl iodides and bromides with tetraethoxysilanes to form aryl silanes such as (4-bromophenyl)triethoxysilane and 4-(triethoxysilyl)acetophenone.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingping Tang et al.
Tetrahedron, 67(24), 4449-4454 (2011-06-22)
A regiospecific silver-mediated fluorination of aryl silanes is reported. The reaction is operationally simple, and employs Ag(2)O as readily available, inexpensive silver source, which can be recovered.
Periodic mesoporous organosilica containing ionic bis-aryl-imidazolium entities: Heterogeneous precursors for silica-hybrid-supported NHC complexes.
Nguyen TP, et al.
Journal of Materials Chemistry, 19(24), 4164-4171 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service