Skip to Content
Merck
All Photos(1)

Documents

639184

Sigma-Aldrich

6-Fluoro-3-pyridinylboronic acid

Synonym(s):

2-Fluoro-5-pyridylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BFNO2
CAS Number:
Molecular Weight:
140.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

OB(O)c1ccc(F)nc1

InChI

1S/C5H5BFNO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3,9-10H

InChI key

OJBYZWHAPXIJID-UHFFFAOYSA-N

Application

Used in a synthesis of halohydroxypyridines by hydroxydeboronation with basic hydrogen peroxide.

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 61, 1417-1417 (2005)

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service