Skip to Content
Merck
All Photos(1)

Key Documents

638943

Sigma-Aldrich

L-(+)-2-Chlorophenylglycine

95%

Synonym(s):

L-2-Chlorophenylglycine, (S)-α−Amino-(2-chlorophenyl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(NH2)CO2H
CAS Number:
Molecular Weight:
185.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

95%

form

solid

optical activity

[α]/D 79.0 to 90.0°, c = 1 in methanol
[α]20/D +79°, c = 1% in chloroform

mp

182-187 °C (lit.)

application(s)

peptide synthesis

SMILES string

N[C@H](C(O)=O)c1ccccc1Cl

InChI

1S/C8H8ClNO2/c9-6-4-2-1-3-5(6)7(10)8(11)12/h1-4,7H,10H2,(H,11,12)/t7-/m0/s1

InChI key

LMIZLNPFTRQPSF-ZETCQYMHSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Xin et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 718305-718305 (2013-06-06)
Coronary heart disease (CHD) is one of the highest mortality diseases in the world. Traditional Chinese medicine compound Danshen dripping pills (CDDPs) have currently made a great achievement in treating CHD. However, the therapeutic mechanism of CDDP is often poorly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service