Skip to Content
Merck
All Photos(1)

Key Documents

637432

Sigma-Aldrich

3,4-Propylenedioxypyrrole-2,5-dicarboxylic acid hydrate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO6 · xH2O
CAS Number:
Molecular Weight:
227.17 (anhydrous basis)
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

solid

SMILES string

O.OC(=O)c1[nH]c(C(O)=O)c2OCCCOc12

InChI

1S/C9H9NO6.H2O/c11-8(12)4-6-7(5(10-4)9(13)14)16-3-1-2-15-6;/h10H,1-3H2,(H,11,12)(H,13,14);1H2

InChI key

AHWAGXGPDRWHBF-UHFFFAOYSA-N

Application

Starting monomer for a variety of propylenedioxypyrrole (ProDOP) conductive and electrooptical polymers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schottland, P.; Reynolds, J.;
Macromolecules, 33, 7051-7051 (2000)
K Zong et al.
The Journal of organic chemistry, 66(21), 6873-6882 (2001-10-13)
New functionalized derivatives of 3,4-ethylenedioxypyrrole (EDOP, 5a) and 3,4-(1,3-propylenedioxy)pyrrole (ProDOP, 5b) as especially electron-rich monomers which yield highly electroactive and stable conducting polymers useful for a diverse set of applications have been synthesized. N-Alkylations of ProDOP were carried out to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service