Skip to Content
Merck
All Photos(1)

Key Documents

636436

Sigma-Aldrich

Methyl 5-aminosalicylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3
CAS Number:
Molecular Weight:
167.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

95-99 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc(N)ccc1O

InChI

1S/C8H9NO3/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,10H,9H2,1H3

InChI key

MXUHMQZOATZRIK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Xiong et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 13(3), 265-267 (1992-05-01)
Methyl 5-aminosalicylate hydrochloride (M-5-AS) at the dosages of 21, 42, 209, and 417 mg.kg-1.d-1 ig to mice during d 6-15 of pregnancy, no obvious effects on the placenta, fetus weight, sex differentiation, external appearance, visceral, and skeletal development were observed.
Y Z Liao et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 11(1), 54-56 (1990-01-01)
Using the improved Kirsner model of ulcerative colitis in rabbits, we found that the therapeutic effect of methyl 5-amino salicylate hydrochloride (M-5-AS) was similar to that of sulfasalazine (salicylazosulfapyridine, SZ). The length of pathological damage and the range of diffuse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service