Skip to Content
Merck
All Photos(1)

Key Documents

636231

Sigma-Aldrich

Isoxazole-5-carbonyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€80.50

€80.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€80.50

About This Item

Empirical Formula (Hill Notation):
C4H2ClNO2
CAS Number:
Molecular Weight:
131.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€80.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.4980 (lit.)

bp

167-168 °C (lit.)

density

1.362 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)c1ccno1

InChI

1S/C4H2ClNO2/c5-4(7)3-1-2-6-8-3/h1-2H

InChI key

NASLINFISOTVJJ-UHFFFAOYSA-N

Application

Isoxazole-5-carbonyl chloride can undergo reaction with 1-benzhydrylpiperazine to give the corresponding 1-benzhydrylpiperazine derivative, which may be used in the treatment of cancer.[1] It can also be used in the synthesis of N-Isoxazole-5-carbonyl-1,3-diphenyl-5-amino-1H-pyrazole, which can act like potent agent in the treatment of neurological and psychiatric illness.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis and in vitro antiproliferative activity of novel 1-benzhydrylpiperazine derivatives against human cancer cell lines"
Kumar A.S.C, et al.
European Journal of Medicinal Chemistry, 44(03), 1223- 1229 (2009)
"An efficient one-pot synthesis of N-(1, 3-diphenyl-1H-pyrazol-5-yl) amides"
Su N-W, et al.
Journal of Heterocyclic Chemistry, 47(04), 831-837 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service