Skip to Content
Merck
All Photos(2)

Key Documents

634549

Sigma-Aldrich

6-Hydroxy-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O2
CAS Number:
Molecular Weight:
148.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

154-158 °C (lit.)

SMILES string

Oc1ccc2CCC(=O)c2c1

InChI

1S/C9H8O2/c10-7-3-1-6-2-4-9(11)8(6)5-7/h1,3,5,10H,2,4H2

InChI key

MOANRQDXNNXOLW-UHFFFAOYSA-N

Application

6-Hydroxy-1-indanone can be used in the synthesis of tetracyclic ketones intermediates during the total syntheses of (±)-frondosin C and (±)-8-epi-frondosin C. It can also be used in the synthesis of 1,4-dihydroindeno[1,2-c]pyrazoles, which can act like potent CHK-1 inhibitors, thereby inhibiting the growth of cancer cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Total syntheses of (?)-frondosin C and (?)-8-epi-frondosin C via a tandem anionic 5-exo dig cyclization?Claisen rearrangement sequence"
Li X. et al.
Tetrahedron, 63(09), 1899-1906 (2007)
"Cyanopyridyl containing 1, 4-dihydroindeno [1, 2-c] pyrazoles as potent checkpoint kinase 1 inhibitors: Improving oral biovailability"
Tong Y, et al.
Bioorganic & Medicinal Chemistry, 17(20), 5665-5670 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service