Skip to Content
Merck
All Photos(1)

Key Documents

633860

2-Chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane-2-d

98 atom % D

Synonym(s):

Enflurane-2-d1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3HClDF5O
CAS Number:
Molecular Weight:
185.50
MDL number:
Pricing and availability is not currently available.

isotopic purity

98 atom % D

mol wt

185.48 by atom % calculation

bp

57 °C (lit.)

density

1.525 g/mL at 25 °C

SMILES string

[2H]C(F)(Cl)C(F)(F)OC(F)F


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service