Skip to Content
Merck
All Photos(1)

Key Documents

633232

Sigma-Aldrich

Acetone-2-13C,d6

99 atom % 13C, 98 atom % D

Synonym(s):

2-Propanone-2-13C-1,1,1,3,3,3-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD313COCD3
CAS Number:
Molecular Weight:
65.11
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % D

Assay

99% (CP)

bp

56 °C (lit.)

mp

-94 °C (lit.)

density

0.885 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H]C([2H])([2H])[13C](=O)C([2H])([2H])[2H]

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3,3+1

InChI key

CSCPPACGZOOCGX-ILPQSMFLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service