Skip to Content
Merck
All Photos(3)

Key Documents

632627

Sigma-Aldrich

Boronic acid, polymer-bound

Boronic acid, polymer-bound

200-400 mesh, extent of labeling: 1.3-2.3 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

polymer bound boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€109.00
5 G
€429.00

€109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€109.00
5 G
€429.00

About This Item

UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€109.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

extent of labeling

1.3-2.3 mmol/g loading

particle size

200-400 mesh

Application

Polymer reagent for oligosaccharide synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Your Boronic acid, polymer bound, cat no 632627 has divinylbenzene crosslinking, but is the polymer backbone a polystyrene?

    1 answer
    1. The polymer is a polystyrene backbone and is crosslinked 1% with divinylbenzene.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service