Skip to Content
Merck
All Photos(1)

Key Documents

632104

Sigma-Aldrich

1,3-Diethynylbenzene

97%

Synonym(s):

1,3-Bis(ethynyl)benzene, m-Diethynylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€35.90
5 G
€173.00

€35.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€35.90
5 G
€173.00

About This Item

Empirical Formula (Hill Notation):
C10H6
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€35.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5820 (lit.)

bp

188-189 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C#Cc1cccc(c1)C#C

InChI

1S/C10H6/c1-3-9-6-5-7-10(4-2)8-9/h1-2,5-8H

InChI key

PNXLPYYXCOXPBM-UHFFFAOYSA-N

Application

1,3-Diethynylbenzene may be used to synthesize mono- and di- 1,4-substituted-1,2,3-triazole ligands, which have the property to self-assemble into metallo-cyclic structures in the presence of silver ions.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Self-assembly of silver (I) metallomacrocycles using unsupported 1, 4-substituted-1, 2, 3-triazole ?click? ligands"
Gower L.M, et al.
Dalton Transactions, 39(9), 2371- 2378 (2010)
Srinivasa R Tala et al.
ACS chemical neuroscience, 9(5), 1001-1013 (2017-12-20)
The melanocortin system is involved in the regulation of complex physiological functions, including energy and weight homeostasis, feeding behavior, inflammation, sexual function, pigmentation, and exocrine gland function. The five melanocortin receptors that belong to the superfamily of G protein-coupled receptors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service