Skip to Content
Merck
All Photos(1)

Key Documents

630829

Sigma-Aldrich

2-Hexyn-1-ol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€56.90

€56.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€56.90

About This Item

Empirical Formula (Hill Notation):
C6H10O
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€56.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.4536 (lit.)

bp

66-67 °C/10 mmHg (lit.)

density

0.893 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCC#CCO

InChI

1S/C6H10O/c1-2-3-4-5-6-7/h7H,2-3,6H2,1H3

InChI key

BTDWSZJDLLLTMI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigeru Tamogami et al.
FEBS letters, 589(3), 390-395 (2015-01-13)
The medicinal herbal plant Achyranthes bidentata (A. bidentata) produces the sweet-odor ester - methyl (E)-2-hexenoate (1) as the major volatile in response to methyl jasmonate (MeJA). Here, we investigated the biosynthetic pathway of methyl (E)-2-hexenoate (1). The common plant precursor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service