Skip to Content
Merck
All Photos(1)

Key Documents

630748

Sigma-Aldrich

7,8-Dihydroxy-4-methylcoumarin

97%

Synonym(s):

4-Methyldaphnetin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€69.30

€69.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€69.30

About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€69.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

242-246 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

CC1=CC(=O)Oc2c(O)c(O)ccc12

InChI

1S/C10H8O4/c1-5-4-8(12)14-10-6(5)2-3-7(11)9(10)13/h2-4,11,13H,1H3

InChI key

NWQBYMPNIJXFNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Can be used as a fluorescent sensor to monitor the consumption of a boronic acid in a Suzuki cross-coupling reaction. Fluorescence is easily detectable to the naked eye using a standard hand-held long-wave UV lamp (365 nm)[1].

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Timothy E Barder et al.
Organic letters, 9(1), 137-139 (2006-12-29)
[reaction: see text] Although boronic acids are widely used in metal-catalyzed reactions, it is difficult to assay their consumption. As such, we developed a reversible fluorescent sensor that is activated upon binding a boronic acid. The sensor can be used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service