Skip to Content
Merck
All Photos(1)

Documents

630659

Sigma-Aldrich

Allyl phenyl carbonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4990 (lit.)

bp

67-70 °C/0.3 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

C=CCOC(=O)Oc1ccccc1

InChI

1S/C10H10O3/c1-2-8-12-10(11)13-9-6-4-3-5-7-9/h2-7H,1,8H2

InChI key

ORUWSEKEVGOAQR-UHFFFAOYSA-N

General description

Allyl phenyl carbonate can be obtained via by reacting phenyl chloroformate and allyl alcohol in presence of pyridine.

Application

Allyl phenyl carbonate can react with (2-alkynyl)phenylisocyanate, catalyzed P(0)/Cu(I) system to give the corresponding indole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"A New Pd0?CuI Bimetallic Catalyst for the Synthesis of Indoles from Isocyanates and Allyl Carbonates"
Kamijo S and Yamamoto Y
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 114(17), 3364-3367 (2002)
?Selective synthesis of carbamate protected polyamines using alkyl phenyl carbonates"
Pittelkow M, et al.
Synthesis, 2002(15), 2195- 2202 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service