Skip to Content
Merck
All Photos(1)

Documents

630438

Sigma-Aldrich

Triethoxy[4-(trifluoromethyl)phenyl]silane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4Si(OC2H5)2
CAS Number:
Molecular Weight:
308.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.436 (lit.)

bp

90-91 °C/0.2 mmHg (lit.)

density

1.098 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)c1ccc(cc1)C(F)(F)F

InChI

1S/C13H19F3O3Si/c1-4-17-20(18-5-2,19-6-3)12-9-7-11(8-10-12)13(14,15)16/h7-10H,4-6H2,1-3H3

InChI key

PBTDWUVVCOLMFE-UHFFFAOYSA-N

Application

A silicon-based nucleophile shown to be reactive in Pd-catalyzed cross-coupling reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service