Skip to Content
Merck
All Photos(1)

Documents

616044

Sigma-Aldrich

Fmoc-Ala-OH-2,3,3,3-d4

98 atom % D

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-alanine-2,3,3,3-d4, L-Alanine-2,3,3,3-d4, N-Fmoc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD(NH-Fmoc)CO2H
Molecular Weight:
315.36
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

optical activity

[α]20/D -18°, c = 1 in DMF

mp

147-153 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+4

SMILES string

[2H]C([2H])([2H])[C@]([2H])(NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m0/s1/i1D3,11D

InChI key

QWXZOFZKSQXPDC-UAMNTACQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle M Koss et al.
Biomacromolecules, 20(1), 422-434 (2018-11-21)
The tissue environment is exceptionally complex, with well-controlled biochemical communication occurring between similar and dissimilar cells as well as between these cells and local extracellular matrices (ECM). To build an artificial ECM that can directly affect regional cell populations, a
K Koss et al.
Journal of controlled release : official journal of the Controlled Release Society, 243, 204-213 (2016-11-05)
(RADA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service