Skip to Content
Merck
All Photos(1)

Key Documents

612731

Supelco

Gadolinium(III) chloride solution

0.1 mg/mL in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
€2,480.00

€2,480.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 EACH
€2,480.00

About This Item

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

€2,480.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

concentration

0.1 mg/mL in D2O (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd/h3*1H;/q;;;+3/p-3

InChI key

MEANOSLIBWSCIT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

1H homogeneity

Quantity

3 mm O.D. tube contains 0.235 mL.
Each pack contains 10 tubes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service