Skip to Content
Merck
All Photos(1)

Documents

611328

Supelco

Iodomethane-13C solution

NMR reference standard, 1% in chloroform-d ("100%", 99.96 atom % D), trimethyl phosphite 1 %, chromium(III) acetylacetonate 0.2 %, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH3I
Molecular Weight:
142.93
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

composition

chromium(III) acetylacetonate, 0.2%
trimethyl phosphite, 1%

concentration

1% in chloroform-d ("100%", 99.96 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

format

single component solution

mass shift

M+1

SMILES string

[13CH3]I

InChI

1S/CH3I/c1-2/h1H3/i1+1

InChI key

INQOMBQAUSQDDS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Indirect detection test

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service