Skip to Content
Merck
All Photos(1)

Documents

609900

Sigma-Aldrich

N-Methylformamide-18O

95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC18ONH(CH3)
CAS Number:
Molecular Weight:
61.07
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

95 atom % 18O

bp

198-199 °C

mp

-4 °C

density

1.044 g/mL at 25 °C

mass shift

M+2

SMILES string

CNC=[18O]

InChI

1S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)/i4+2

InChI key

ATHHXGZTWNVVOU-DOMIDYPGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Moreno et al.
Current biology : CB, 29(1), 23-34 (2018-12-18)
The plasticity of developing tissues relies on the adjustment of cell survival and growth rate to environmental cues. This includes the effect of mechanical cues on cell survival. Accordingly, compaction of an epithelium can lead to cell extrusion and cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service