Skip to Content
Merck
All Photos(1)

Documents

608645

Sigma-Aldrich

Tamoxifen-(N,N-dimethyl-13C2)-15N

99 atom % 13C, 99 atom % 15N, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C24H2915NO
Molecular Weight:
374.49
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

isotopic purity

99 atom % 13C
99 atom % 15N

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

SMILES string

CC\C(c1ccccc1)=C(\c2ccccc2)c3ccc(OCC[15N]([13CH3])[13CH3])cc3

InChI

1S/C26H29NO/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3/h5-18H,4,19-20H2,1-3H3/b26-25-/i2+1,3+1,27+1

InChI key

NKANXQFJJICGDU-MEAUAUGVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service