Skip to Content
Merck
All Photos(1)

Documents

606065

Sigma-Aldrich

L-Lactic acid-13C3 solution

greener alternative

85 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3H6O3
CAS Number:
Molecular Weight:
93.06
MDL number:
UNSPSC Code:
12352106

description

Packaged as gram of solution

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)
≥98% (Chiral Purity, HPLC)

greener alternative product score

old score: 45
new score: 14
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

85 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

greener alternative category

mass shift

M+3

storage temp.

−20°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses”, “Design for Energy Efficiency” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service