Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
form
solid
optical activity
[α]25/D +141°, c = 0.4 in acidified ammonium molybdate
mp
167-170 °C (lit.)
mass shift
M+6
SMILES string
O[13CH2][13C@@H](O)[13C@@H](O)[13C@H](O)[13C@H](O)[13CH2]O
InChI
1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
InChI key
FBPFZTCFMRRESA-CRWJSTOYSA-N
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service