All Photos(1)
About This Item
Linear Formula:
BrC6H2(OC2H5)(CH3)B(OH)2
CAS Number:
Molecular Weight:
258.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
mp
98-102 °C (lit.)
SMILES string
CCOc1c(Br)cc(C)cc1B(O)O
InChI
1S/C9H12BBrO3/c1-3-14-9-7(10(12)13)4-6(2)5-8(9)11/h4-5,12-13H,3H2,1-2H3
InChI key
BTBLZNZUELXCOT-UHFFFAOYSA-N
Other Notes
Contains varying amounts of anhydride
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service