Skip to Content
Merck
All Photos(1)

Key Documents

595829

Sigma-Aldrich

Nicotinic acid-13C6

99 atom % 13C, 99% (CP)

Synonym(s):

Niacin-13C6, Vitamin B3-13C6

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€2,560.00

€2,560.00


Check Cart for Availability


Select a Size

Change View
10 MG
€2,560.00

About This Item

Empirical Formula (Hill Notation):
13C6H5NO2
CAS Number:
Molecular Weight:
129.07
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.13

€2,560.00


Check Cart for Availability

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

236-239 °C (lit.)

mass shift

M+6

SMILES string

O[13C](=O)[13c]1[13cH][13cH][13cH]n[13cH]1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

PVNIIMVLHYAWGP-IDEBNGHGSA-N

Related Categories

General description

Nicotinic acid (pyridine-3-carboxylic acid) is also known as niacin or vitamin B3. It is a water-soluble vitamin found in some vegetable and animal sources. Niacin is also a precursor for the synthesis of NAD and NADP.[1][2]

Application

Nicotinic acid-13C6 is also known as niacin-13C6. This product can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Niacin, Nicotinamide, and Nicotinic Acid
Van A and Susan D
Kirk-Othmer Encyclopedia of Chemical Technology, 349, 4379-4379 (2000)
Effect on cardiovascular risk of high density lipoprotein targeted drug treatments niacin, fibrates, and CETP inhibitors: meta-analysis of randomised controlled trials including 117 411 patients
Keene D, et al.
BMJ (Clinical Research ed.), 349, 4379-4379 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service