Skip to Content
Merck
All Photos(2)

Key Documents

593400

Sigma-Aldrich

3-Chloro-5-fluorosalicylaldehyde

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€78.90

€78.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€78.90

About This Item

Linear Formula:
ClC6H2(F)(OH)CHO
CAS Number:
Molecular Weight:
174.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€78.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

95-99 °C (lit.)

functional group

aldehyde
chloro
fluoro

SMILES string

Oc1c(Cl)cc(F)cc1C=O

InChI

1S/C7H4ClFO2/c8-6-2-5(9)1-4(3-10)7(6)11/h1-3,11H

InChI key

VHXIEGGPMOEWNK-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service