Skip to Content
Merck
All Photos(3)

Key Documents

593168

Sigma-Aldrich

Ethyl 3,4-dimethoxybenzoylacetate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€161.00

About This Item

Linear Formula:
C2H5OCOCH2COC6H3(OCH3)2
CAS Number:
Molecular Weight:
252.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€161.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

37-41 °C (lit.)

functional group

ester
ketone

SMILES string

CCOC(=O)CC(=O)c1ccc(OC)c(OC)c1

InChI

1S/C13H16O5/c1-4-18-13(15)8-10(14)9-5-6-11(16-2)12(7-9)17-3/h5-7H,4,8H2,1-3H3

InChI key

BHSYONGONOKNAI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenzo Vannozzi et al.
International journal of nanomedicine, 11, 69-91 (2016-01-06)
This work aims to demonstrate the possibility to fabricate ultra-thin polymeric films loaded with an anti-restenotic drug and capable of tunable drug release kinetics for the local treatment of restenosis. Vascular nanopatches are composed of a poly(lactic acid) supporting membrane
Vito Coviello et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 117, 276-285 (2017-05-01)
Nano-sized colloidal carriers represent innovative drug delivery systems, as they allow a targeted and prolonged release of poorly water-soluble drugs, improving their bioavailability and modifying their pharmacokinetic parameters. In this work we describe cyclodextrin-based nanosponges, obtained through polimerization of β-cyclodextrin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service