Skip to Content
Merck
All Photos(1)

Key Documents

592161

Sigma-Aldrich

1-Bromo-3,5-di-tert-butylbenzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€27.70
10 G
€251.00

€27.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€27.70
10 G
€251.00

About This Item

Linear Formula:
BrC6H3(C(CH3)3)2
CAS Number:
Molecular Weight:
269.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€27.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

62-66 °C (lit.)

functional group

bromo

SMILES string

CC(C)(C)c1cc(Br)cc(c1)C(C)(C)C

InChI

1S/C14H21Br/c1-13(2,3)10-7-11(14(4,5)6)9-12(15)8-10/h7-9H,1-6H3

InChI key

BUOWTUULDKULFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter C Ho et al.
Dalton transactions (Cambridge, England : 2003), 48(15), 4879-4886 (2019-03-21)
Pt(ii) and Rh(iii) readily form complexes with 3-methyl-5-aryl-1,2-tellurazole 2-oxides (L = (3-Me-5-Ar-1,2-C3HTe(NO)). The aryl group is either phenyl or 3,5-di-tert-butylphenyl, the latter case being a new derivative with enhanced solubility. The compound [Pt(3-Me-5-(3,5-tBu2C6H3)-1,2-C3HTe(NO))4](BF4)2 crystallizes in the P21/C space group featuring

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service