Skip to Content
Merck
All Photos(1)

Documents

591963

Sigma-Aldrich

Poly(2,2,2-trifluoroethyl methacrylate)

Synonym(s):

2,2,2-Trifluoroethyl methacrylate polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH2CF3)]n
CAS Number:
MDL number:
UNSPSC Code:
12352103

form

solid

refractive index

n20/D 1.418 (lit.)
n20/D 1.42 (lit.)

viscosity

12.6 cP, 1 % in methylene chloride

mp

50-52 °C (lit.)

transition temp

Tg 69 °C(lit.)

solubility

DMSO: soluble
NMP: soluble
THF: soluble
methylene chloride: soluble

InChI

1S/C6H7F3O2/c1-4(2)5(10)11-3-6(7,8)9/h1,3H2,2H3

InChI key

QTKPMCIBUROOGY-UHFFFAOYSA-N

Application

Low refractive index polymer for cladding layer in optical waveguides. Transparent, low surface tension polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service