Skip to Content
Merck
All Photos(1)

Key Documents

591386

Sigma-Aldrich

Tridecanoic-2,2-d2 acid

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CD2CO2H
CAS Number:
Molecular Weight:
216.36
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

form

solid

bp

236 °C/100 mmHg

mp

41-42 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(CCCCCCCCCCC)C(O)=O

InChI

1S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)/i12D2

InChI key

SZHOJFHSIKHZHA-XUWBISKJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satya Girish Chandra Avula et al.
Journal of separation science, 40(10), 2214-2227 (2017-03-23)
Algae can synthesize, accumulate and store large amounts of lipids in its cells, which holds immense potential as a renewable source of biodiesel. In this work, we have developed and validated a GC-MS method for quantitation of fatty acids and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service